Learn More
Cole-Parmer Cole-Parmer VapLock™ Waste Funnel, 4" dia., Justrite Centura, SS / PTFE quick disconnect, HDPE; 1/ea
Catalog No. NC1496256 Shop All Cole-Parmer Products
Encompass
Use these 4" diameter funnels to safely dispose of liquid laboratory waste. A locking hinged lid minimizes leaks and spills when the funnel is closed. Quick-disconnect fitting on models 12020-64 and -65 allows fast and easy connection to Justrite Centura safety containers. Funnels are constructed of high-density polyethylene (HDPE) and polypropylene (PP) with a Viton gasket.
Catalog No. NC1496256 Supplier Cole-Parmer Supplier No. 1202065
Use these 4" diameter funnels to safely dispose of liquid laboratory waste. A locking hinged lid minimizes leaks and spills when the funnel is closed. Quick-disconnect fitting on models 12020-64 and -65 allows fast and easy connection to Justrite Centura safety containers. Funnels are constructed of high-density polyethylene (HDPE) and polypropylene (PP) with a Viton gasket.