Learn More
Cole-Parmer™ VapLock™ Waste Funnels
Catalog No. 13311266
Change view
Click to view available options
Material:
HDPE, Polypropylene
Polypropylene and EPDM quick disconnect, HDPE funnel
SS and PTFE quick disconnect, HDPE funnel
For Use With (Equipment):
Container Ports with 1/2 in. NPT(F)
Justrite Centura Containers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material For Use With (Equipment)
13-311-266 Polypropylene and EPDM quick disconnect, HDPE funnel Justrite Centura Containers
13-311-267 SS and PTFE quick disconnect, HDPE funnel Justrite Centura Containers
13-311-268 HDPE, Polypropylene Container Ports with 1/2 in. NPT(F)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-311-266 Supplier Cole-Parmer™ Supplier No. 1202064
Add to Cart
Add to Cart
VapLock™ Waste Funnels safely and securely capture waste and hazardous solvents
  • Reduces fumes and harmful vapor exhaust
  • Easily connect to Justrite™ Centura safety containers
Warranty and Services
None
Specifications
For Use With (Equipment) Justrite Centura Containers
Material Polypropylene and EPDM quick disconnect, HDPE funnel
For Use With (Application) Solvent Safety
Type Waste Funnel, 4 in. dia