Learn More
Synthware™ Vapor Duct
GREENER_CHOICESDP
Catalog No. 31500628 Shop All Kemtech Products
Change view
Click to view available options
Product Type:
Labtech-style Vapor Duct
Catalog No. Product Type
31-500-628 Labtech-style Vapor Duct
1 options
Catalog No. 31-500-628 Supplier Synthware™ Supplier No. V222440L
Add to Cart
Add to Cart
Precision ground and polished.
Specifications
Product Type Labtech-style Vapor Duct
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Standard Taper Joint 24/40
Quantity 1 Ea.