Learn More
Ace Glass, Inc. Vapor duct tube, 24/40 inner, 55mm, Buchi 32335 & 32341
SDP
Catalog No. NC9693496
Encompass
45mm long replacement vapor duct tube for Buchi R-114/124/134/144 rotary evaporator V assemblies. End joints are 24/40 [ST] inner joints, middle section has GL thread to accomm ODate a Combi-Clip, and the other end is a straight tube.
Catalog No. NC9693496 Supplier Ace Glass, Inc. Supplier No. 1328528
45mm long replacement vapor duct tube for Buchi R-114/124/134/144 rotary evaporator V assemblies. End joints are 24/40 [ST] inner joints, middle section has GL thread to accomm ODate a Combi-Clip, and the other end is a straight tube.