Learn More
Description
- NIOSH-approved protection against Mercury Vapor, Chlorine, Sulfur Dioxide, Organic Vapor and Hydrogen Sulfide
- NIOSH-approved P100 particulate filters provide respiratory protection against certain oil and non-oil based particulates
- Particulate filter is permanently attahed to cartridge for easy, one-step assembly
- Bayonet connection is compatibile with a variety of 3M™ respirators
- Swept back design enhances field of view and comfort
- Wide application range reduces the need for large inventories
- For workplace and occupational use only
Specifications
Specifications
| Cartridge Type | Mercury Vapor, Organic Vapor, Acid Gas Cartridge |
| Connection Type | Bayonet |
| Color | Olive/Magenta |
| Filter Type | P100 |
| Product Type | Cartridge/Filter Combination |
| Filter Efficiency | 100 |
| For Use With (Application) | Laboratories, Line Opening, Chemical Transfer |
| For Use With (Equipment) | 3MTM Full Facepieces 6000, 7800, FF-400 Series; Half Facepiece 6000, 6500 7500 Series; 3MTM ScottTM AV-3000 HT Facepieces, 3MTM ScottTM AV-3000 SureSeal Facepieces |
| Model | 60927 |
| Quantity | 30 Pairs/Cs. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.