Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Mercury Vapor/Organic Vapor/Acid Gas Cartridges 6007

Catalog No. 19072197 Shop All 3M Products
Change view
Click to view available options
Bayonet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-072-197 Bayonet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-197 Supplier 3M™ Supplier No. 7100198706
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NIOSH-approved cartridge for protection against mercury vapor, chlorine, sulfur dioxide, organic vapor and hydrogen sulfide.

  • NIOSH-approved protection against Mercury Vapor, Chlorine, Sulfur Dioxide, Organic Vapor and Hydrogen Sulfide
  • Bayonet connection is compatibile with a variety of 3M™ respirators
  • Swept back design provides enhanced field of view and comfort
  • Wide application range reduces the need for large inventories
  • For workplace and occupational use only
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Vapor, Gas
Connection Type Bayonet
Color Olive
Product Type Cartridge
For Use With (Application) Chemical Transfer, Laboratories, Line Opening
For Use With (Equipment) 3MTM Full Facepieces 6000, 7800, FF-400 Series; Half Facepiece Respirators 6000, 6500, 7500 Series; 3MTM ScottTM AV-3000 HT Facepieces, 3MTM ScottTM AV-3000 SureSeal Facepieces
Model 6007
Quantity 30 Pairs/Cs.
Length (Metric) 107 mm
Height (Metric) 86 mm
Weight (Metric) 93 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.