Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Vapor Pressure Chemistry Demo Kit
GREENER_CHOICESCI_ED

Catalog No. S37755 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S37755 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
DOT: Non regulated
Catalog No. S37755 Supplier Innovating Science™ Supplier No. IS8015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOT: Non regulated

Includes

2 x 2.5kg Sodium Chloride Crystals

Proves Raoult's Law empirically by measuring the change in vapor pressure as solute is added to a solvent.

  • Measurements are taken at different temperatures to demonstrate effect of temperature on vapor pressure

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Proves Raoult Law empirically by measuring the change in vapor pressure as solute is added to a solvent.
Includes 2 x 2.5kg Sodium Chloride Crystals
Quantity 1
Dimensions (L x W x H) 10 x 7 x 12.375 in.
Class Size 15 Lab Groups
Product Type Vapor Pressure Chemistry Demo Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.