Learn More
Kem Medical Products Corp VAPOR TRAK FORMALDEHYDE 4/PK
SDP
Catalog No. NC9150889
Encompass_Preferred
Formaldehyde monitor has been validated for both the OSHA 8 hr. TWA exposure limit of 0.75ppm (0.94mg/m3) and the 15 min. STEL limit of 2.0ppm
Catalog No. NC9150889 Supplier Kem Medical Products Corp Supplier No. 8510
Formaldehyde monitor has been validated for both the OSHA 8 hr. TWA exposure limit of 0.75ppm (0.94mg/m3) and the 15 min. STEL limit of 2.0ppm