Learn More
Chemglass Life Sciences Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 173mm OAL, Decaled "R111/140" Length A is 45mm
SDP
Catalog No. NC0175974
Encompass_Preferred
Decaled "R111/140" Length A is 45mm. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. NOT SUPPLIED WITH COMBI-CLIP. Equal to Buchi Part # 23738. Buchi is a registered trade mark of B¿HI Labortechnik AG.
Catalog No. NC0175974 Supplier Chemglass Life Sciences Supplier No. CG1317G12
Decaled "R111/140" Length A is 45mm. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. NOT SUPPLIED WITH COMBI-CLIP. Equal to Buchi Part # 23738. Buchi is a registered trade mark of B¿HI Labortechnik AG.