Learn More
Chemglass Life Sciences Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 288mm OAL, Decaled "R210/215", for Glass Assembly A, Length "A" = 152mm, 288mm OAL
SDP
Catalog No. 501531072
Encompass_Preferred
Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 288mm OAL, Decaled "R210/215", for Glass Assembly A, Length "A" = 152mm, 288mm OAL. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. Available in the listed sizes. NOT SUPPLIED WITH COMBI-CLIP.
Catalog No. 50-153-1072 Supplier Chemglass Life Sciences Supplier No. CG1317G18
Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 288mm OAL, Decaled "R210/215", for Glass Assembly A, Length "A" = 152mm, 288mm OAL. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. Available in the listed sizes. NOT SUPPLIED WITH COMBI-CLIP.