Learn More
Chemglass Life Sciences Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 288mm OAL, Decaled "R111/140", Length A is 160mm
SDP
Catalog No. 501531071
Encompass_Preferred
Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 288mm OAL, Decaled "R111/140", Length A is 160mm. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip.
Catalog No. 50-153-1071 Supplier Chemglass Life Sciences Supplier No. CG1317G10
Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 288mm OAL, Decaled "R111/140", Length A is 160mm. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip.