Description
- Provides a snug connection from the oxygen source to the inlet of the vaporizer
- Provides a snug connection from the outlet of the vaporizer to the anesthesia system
- The ¼ in. hose barb accepts 3/16 in. ID silicone tubing
- Three feet of 3/16 in. ID silicone tubing included
Specifications
Specifications
| Autoclavable | Autoclavable |
| Animal | All Animals |
| Type | Tube Kit |
| Disposable | Reusable |
| Size | Standard |
| Sterility | Non-sterile |