Learn More
SOMNI Scientific™ Vaporizer Service Exchange Draeger 2000 Iso (Specify Fill Type, Mount)
SDP
Catalog No. 04777160 Shop All Somni Scientific Products
Change view
Click to view available options
Model:
Draeger 2000 Isoflurane
Catalog No. Model
04-777-160 Draeger 2000 Isoflurane
1 options
Catalog No. 04-777-160 Supplier SOMNI Scientific™ Supplier No. VX6023
Only null left
Add to Cart
Add to Cart
Maintains device accuracy, proper function and prevents exposure from leaks. A fully serviced and calibrated vaporizer of the same type is shipped to the customer. Customer uses prepaid return label to send their vaporizer back to our service center. No downtime.
  • Isoflurane, Draeger 2000
  • Specify Fill type, Mount and Serial number when ordering
  • Maintains accuracy
  • Prevents exposure from leaks
  • Full Service and Calibration
    • Certificate of Calibration included
  • Service interval based on manufacturer recommendation
  • Warranty provided
Specifications
For Use With (Application) Animal Inhalant Anesthesia
For Use With (Equipment) RM-17003, RM-17004, AMD-17004, TT-17000, OC-21003, OC-21004
Material Brass
Length (Metric) 18.8 cm
Width (Metric) 10.8 cm
Model Draeger 2000 Isoflurane
Includes Calibration Certificate
For Veterinary Use Only