Learn More
Alkali Scientific VaporSeal™ Gas Permeable Membrane, Hydrophobic Sealing Film
SDP
Catalog No. 502388959
Encompass_Preferred
VaporSeal™ features a Hydrophobic Sealing Film, gas permeable membrane with a medical grade, non-cytotoxic adhesive. The porous membrane consists of layers of heat welded rayon fibers. Moisture vapor transmission rate = 4,200g/m2/24hr. Easy-to-pierce with serological pipets and pipet tips for direct sample recovery.
Catalog No. 50-238-8959 Supplier Alkali Scientific Supplier No. SF1VP
VaporSeal™ features a Hydrophobic Sealing Film, gas permeable membrane with a medical grade, non-cytotoxic adhesive. The porous membrane consists of layers of heat welded rayon fibers. Moisture vapor transmission rate = 4,200g/m2/24hr. Easy-to-pierce with serological pipets and pipet tips for direct sample recovery.