Learn More
Alkali Scientific VaporSeal™ Titan™ Microplate Sealing Rolls, 2 Rolls/pk, 50 Sheets/Roll
SDP
Catalog No. 502388905
Encompass_Preferred
The dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Environmentally friendly; no paper liner to remove and discard; utilizes approximately 59% less material than AeraSeal sheeted film. Each sheet is separated by an adhesive-free, perforated, color-coded red band. The smooth lower surface of the dispenser can be used to seal the film to the plate. Dispenser protects unused seals from contamination. Lower support prop prevents dispensing bill from contacting surfaces to prevent contamination. Replacement rolls can be easily installed by releasing the locking clasps on the dispenser. Moisture vapor transmission rate = 4,200g/m²/24hr. (The dispenser sold seperately. This item is only replacement rolls. The starter kit catalog number is SF3VP or SF3VP-S for sterile version.)
Catalog No. 50-238-8905 Supplier Alkali Scientific Supplier No. SF2VP
The dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Environmentally friendly; no paper liner to remove and discard; utilizes approximately 59% less material than AeraSeal sheeted film. Each sheet is separated by an adhesive-free, perforated, color-coded red band. The smooth lower surface of the dispenser can be used to seal the film to the plate. Dispenser protects unused seals from contamination. Lower support prop prevents dispensing bill from contacting surfaces to prevent contamination. Replacement rolls can be easily installed by releasing the locking clasps on the dispenser. Moisture vapor transmission rate = 4,200g/m²/24hr. (The dispenser sold seperately. This item is only replacement rolls. The starter kit catalog number is SF3VP or SF3VP-S for sterile version.)