Learn More
Medchemexpress LLC HY-P0061A 5mg Medchemexpress, Vapreotide (acetate) CAS:849479-74-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0061A5MG
Encompass_Preferred
Medchemexpress, HY-P0061A 5mg Vapreotide (acetate) CAS:849479-74-9 Vapreotide acetate (RC-160 acetate; BMY-41606 acetate) is a neurokinin-1 (NK1) receptor antagonist, with an IC50 of 330 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0515