Learn More
Medchemexpress LLC Vapreotide acetate | 849479-74-9 | >98.16% | 1191.42 | 25 MG
SDP
Catalog No. 5000426871
Encompass_Preferred
Vapreotide acetate is a neurokinin-1 (NK1) receptor antagonist, characterized by an IC50 of 330 nM. It attenuates Substance P (SP)-triggered intracellular calcium increases and NF-κB activation, while inhibiting SP-induced IL-8 and MCP-1 production. It also inhibits HIV-1 infection and significantly reduces GH-, PRL, and/or alpha-subunit release from pituitary adenoma cells. The compound exhibits moderate-to-high affinities for SSTR2, -3, and -5.
  • Neurokinin-1 (NK1) receptor antagonist.
  • Attenuates SP-triggered calcium increases, NF-κB activation.
  • Inhibits SP-induced IL-8 and MCP-1 production.
  • Inhibits HIV-1 infection; reduces GH, PRL, and alpha-subunit release.
  • Moderate-to-high affinities for SSTR2, -3, and -5.
  • Can inhibit androgen-independent prostate cancer growth and decrease collateral circulation in cirrhotic rats.
Catalog No. 50-004-26871 Supplier Medchemexpress LLC Supplier No. 25MGHYP0061A
Vapreotide acetate is a neurokinin-1 (NK1) receptor antagonist, characterized by an IC50 of 330 nM. It attenuates Substance P (SP)-triggered intracellular calcium increases and NF-κB activation, while inhibiting SP-induced IL-8 and MCP-1 production. It also inhibits HIV-1 infection and significantly reduces GH-, PRL, and/or alpha-subunit release from pituitary adenoma cells. The compound exhibits moderate-to-high affinities for SSTR2, -3, and -5.
  • Neurokinin-1 (NK1) receptor antagonist.
  • Attenuates SP-triggered calcium increases, NF-κB activation.
  • Inhibits SP-induced IL-8 and MCP-1 production.
  • Inhibits HIV-1 infection; reduces GH, PRL, and alpha-subunit release.
  • Moderate-to-high affinities for SSTR2, -3, and -5.
  • Can inhibit androgen-independent prostate cancer growth and decrease collateral circulation in cirrhotic rats.