Learn More
Medchemexpress LLC Vapreotide acetate | 849479-74-9 | 98.2% | 1191.42 | 10 MG
SDP
Catalog No. 5000426312
Encompass_Preferred
Vapreotide acetate is a neurokinin-1 (NK1) receptor antagonist with an IC50 of 330 nM. It is intended for research use only.
  • Attenuates Substance P (SP)-triggered intracellular calcium increases and NF-κB activation.
  • Inhibits SP-induced IL-8 and MCP-1 production.
  • Inhibits HIV-1 infection of human MDM in vitro.
  • Inhibits GH-, PRL, and/or alpha-subunit release by human GH-secreting pituitary adenoma cells.
  • Exhibits affinities for SSTR2, -3, and -5.
  • Reduces tumor volumes and weights in androgen-independent prostate cancer xenografts.
  • Decreases collateral circulation blood flow in cirrhosis and attenuates its development.
Catalog No. 50-004-26312 Supplier Medchemexpress LLC Supplier No. HYP0061A10MG
Vapreotide acetate is a neurokinin-1 (NK1) receptor antagonist with an IC50 of 330 nM. It is intended for research use only.
  • Attenuates Substance P (SP)-triggered intracellular calcium increases and NF-κB activation.
  • Inhibits SP-induced IL-8 and MCP-1 production.
  • Inhibits HIV-1 infection of human MDM in vitro.
  • Inhibits GH-, PRL, and/or alpha-subunit release by human GH-secreting pituitary adenoma cells.
  • Exhibits affinities for SSTR2, -3, and -5.
  • Reduces tumor volumes and weights in androgen-independent prostate cancer xenografts.
  • Decreases collateral circulation blood flow in cirrhosis and attenuates its development.