Learn More
Medchemexpress LLC Vapreotide acetate | 849479-74-9 | 98.2% | 1191.42 | 50 MG
SDP
Catalog No. 5000425425
Encompass_Preferred
Vapreotide acetate, also known as RC-160 acetate, is a neurokinin-1 (NK1) receptor antagonist. It inhibits Substance P (SP)-triggered intracellular calcium increases and NF-κB activation, making it suitable for various research applications in cell signaling and disease models.
  • Neurokinin-1 (NK1) receptor antagonist with an IC50 of 330 nM
  • Inhibits Substance P (SP)-induced IL-8 and MCP-1 production
  • Demonstrates activity against HIV-1 infection in human MDM in vitro
  • Effective in decreasing collateral circulation blood flow in cirrhosis in vivo
  • Reduces tumor volumes in prostate-cancer xenografts in nude mice
Catalog No. 50-004-25425 Supplier Medchemexpress LLC Supplier No. HYP0061A50MG
Vapreotide acetate, also known as RC-160 acetate, is a neurokinin-1 (NK1) receptor antagonist. It inhibits Substance P (SP)-triggered intracellular calcium increases and NF-κB activation, making it suitable for various research applications in cell signaling and disease models.
  • Neurokinin-1 (NK1) receptor antagonist with an IC50 of 330 nM
  • Inhibits Substance P (SP)-induced IL-8 and MCP-1 production
  • Demonstrates activity against HIV-1 infection in human MDM in vitro
  • Effective in decreasing collateral circulation blood flow in cirrhosis in vivo
  • Reduces tumor volumes in prostate-cancer xenografts in nude mice