Learn More
ELITechGroup VAPRO Vapor Pressure Osmometer
SDP
Catalog No. NC0044806
Encompass
The Vapro Vapor Pressure Osmometer, can be used to determine the osmolality in any type of sample: fluids, viscous, and even tissue, giving the Vapro great versatility in clinical medicine or biomedical research applications. The Vapro osmometer requires a sample volume of only 10 microliters. Osmolality is displayed in 90 seconds. Measurement range: Typically 20 to 3200 mmol/kg* (up to 3600 mmol/kg with extended range osmometer) @ 25C ambient.
Catalog No. NC0044806 Supplier ELITechGroup Supplier No. MODEL 5600
Add to Cart
Add to Cart
Request a Quote
The Vapro Vapor Pressure Osmometer, can be used to determine the osmolality in any type of sample: fluids, viscous, and even tissue, giving the Vapro great versatility in clinical medicine or biomedical research applications. The Vapro osmometer requires a sample volume of only 10 microliters. Osmolality is displayed in 90 seconds. Measurement range: Typically 20 to 3200 mmol/kg* (up to 3600 mmol/kg with extended range osmometer) @ 25C ambient.