Learn More
Chemglass Life Sciences Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 173mm OAL, Decaled "R114/144" Length A is 45mm
SDP
Catalog No. NC1664889
Encompass_Preferred
Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 173mm OAL, Decaled "R114/144" Length A is 45mm. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. NOT SUPPLIED WITH COMBI-CLIP.
Catalog No. NC1664889 Supplier Chemglass Life Sciences Supplier No. CG1317G16
Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 173mm OAL, Decaled "R114/144" Length A is 45mm. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. NOT SUPPLIED WITH COMBI-CLIP.