Learn More
Cole-Parmer Master Appliance VT-751D Variable Temperature Heat Gun 100° to 1200°F, 1740 Watts, 120 V, 60 Hz
Supplier:  Cole-Parmer 0302600
Encompass_Preferred
Heat guns measure 10"L x 5.25"W x 9.5"H; include a 6-ft neoprene-jacketed cord. Order attachments and replacement heating element kits separately.
Catalog No. NC0540779
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov