Learn More
Apexbio Technology LLC Vardenafil,10mg CAS# 224785-90-4
SDP
Supplier:  Apexbio Technology LLC B334310
Encompass_Preferred
Vardenafil is a potent and selective inhibitor of phosphodiesterase type5 (PDE5) with the IC50 value of 0.7 nM in enzymatic assay in vitro[1].Vardenafil has shown a greater selective inhibition towards PDE5 than PDEs from bovine, with the IC50 values of 180nM,>10000nM, 2500nM, 4000nM and 11nM for PDE1, PDE2, PDE3, PDE4 and PDE6, respectively. Besides, Vardenafil has been reported to significantly enhance the SNP-induced, ACh-induced and transmural electrical stimulation-induced relaxation of human trabecular smooth muscle. Vardenafil has been revealed to increase the amount of cGMP in human corpus cavernosum. Apart from these, Vardenafil has also enhanced the length of penile in the conscious rabbit [1]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0307