Learn More
Apexbio Technology LLC Vardenafil 224785-90-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063912
Encompass_Preferred
Vardenafil (CAS 224785-90-4) is a potent and selective inhibitor of phosphodiesterase type 5 (PDE5) It inhibits PDE5 enzyme activity with an IC50 of approximately 0 7 nM in vitro Compared with other PDE isoforms (PDE1-4 and PDE6) vardenafil demonstrates superior selectivity toward PDE5 though moderate inhibition of PDE6 (IC50 11 nM) is observed Vardenafil enhances cGMP accumulation in human corpus cavernosum and significantly potentiates nitric oxide (SNP)- acetylcholine- and electrical field stimulation-induced relaxation in human trabecular smooth muscle Its vasodilatory effects render it suitable for research in erectile function and related vascular physiology
Catalog No. 50-000-63912 Supplier Apexbio Technology LLC Supplier No. B33435.1
 Restricted Item
Add to Cart
Add to Cart
Vardenafil (CAS 224785-90-4) is a potent and selective inhibitor of phosphodiesterase type 5 (PDE5) It inhibits PDE5 enzyme activity with an IC50 of approximately 0 7 nM in vitro Compared with other PDE isoforms (PDE1-4 and PDE6) vardenafil demonstrates superior selectivity toward PDE5 though moderate inhibition of PDE6 (IC50 11 nM) is observed Vardenafil enhances cGMP accumulation in human corpus cavernosum and significantly potentiates nitric oxide (SNP)- acetylcholine- and electrical field stimulation-induced relaxation in human trabecular smooth muscle Its vasodilatory effects render it suitable for research in erectile function and related vascular physiology