Learn More
Selleck Chemical LLC Vardenafil HCl Trihydrate 10mg 330808-88-3 BAY38-9456
SDP
Supplier:  Selleck Chemical LLC S251510MG
Encompass_Preferred
Vardenafil HCl Trihydrate (BAY38-9456) is a new type PDE inhibitor with IC50 of 0.7 and 180 nM for PDE5 and PDE1, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-552-8