Learn More
Apexbio Technology LLC Vardenafil HCl Trihydrate 330808-88-3 10mg
SDP
Catalog No. 5000095589
Encompass_Preferred
Vardenafil HCl Trihydrate (CAS 330808-88-3) is the trihydrate salt form of vardenafil a potent and selective inhibitor of phosphodiesterase type 5 (PDE5) In enzymatic assays vardenafil demonstrates an IC50 of 0 7 nM for PDE5 exhibiting high selectivity over other phosphodiesterase isoforms with significantly higher IC50 values for PDE1 PDE2 PDE3 PDE4 and PDE6 Vardenafil enhances relaxation of human trabecular smooth muscle in response to sodium nitroprusside acetylcholine and electrical stimulation and increases cGMP concentrations in human corpus cavernosum tissue These properties establish its utility in studies addressing cGMP-mediated smooth muscle relaxation and PDE5-related pharmacology
Catalog No. 50-000-95589 Supplier Apexbio Technology LLC Supplier No. A432310
Vardenafil HCl Trihydrate (CAS 330808-88-3) is the trihydrate salt form of vardenafil a potent and selective inhibitor of phosphodiesterase type 5 (PDE5) In enzymatic assays vardenafil demonstrates an IC50 of 0 7 nM for PDE5 exhibiting high selectivity over other phosphodiesterase isoforms with significantly higher IC50 values for PDE1 PDE2 PDE3 PDE4 and PDE6 Vardenafil enhances relaxation of human trabecular smooth muscle in response to sodium nitroprusside acetylcholine and electrical stimulation and increases cGMP concentrations in human corpus cavernosum tissue These properties establish its utility in studies addressing cGMP-mediated smooth muscle relaxation and PDE5-related pharmacology