Learn More
Apexbio Technology LLC Vardenafil HCl Trihydrate 330808-88-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093582
Encompass_Preferred
Vardenafil HCl Trihydrate (CAS 330808-88-3) is the trihydrate salt form of vardenafil a potent and selective inhibitor of phosphodiesterase type 5 (PDE5) In enzymatic assays vardenafil demonstrates an IC50 of 0 7 nM for PDE5 exhibiting high selectivity over other phosphodiesterase isoforms with significantly higher IC50 values for PDE1 PDE2 PDE3 PDE4 and PDE6 Vardenafil enhances relaxation of human trabecular smooth muscle in response to sodium nitroprusside acetylcholine and electrical stimulation and increases cGMP concentrations in human corpus cavernosum tissue These properties establish its utility in studies addressing cGMP-mediated smooth muscle relaxation and PDE5-related pharmacology
Catalog No. 50-000-93582 Supplier Apexbio Technology LLC Supplier No. A43235.1
Vardenafil HCl Trihydrate (CAS 330808-88-3) is the trihydrate salt form of vardenafil a potent and selective inhibitor of phosphodiesterase type 5 (PDE5) In enzymatic assays vardenafil demonstrates an IC50 of 0 7 nM for PDE5 exhibiting high selectivity over other phosphodiesterase isoforms with significantly higher IC50 values for PDE1 PDE2 PDE3 PDE4 and PDE6 Vardenafil enhances relaxation of human trabecular smooth muscle in response to sodium nitroprusside acetylcholine and electrical stimulation and increases cGMP concentrations in human corpus cavernosum tissue These properties establish its utility in studies addressing cGMP-mediated smooth muscle relaxation and PDE5-related pharmacology