Learn More
Medchemexpress LLC HY-B0442A 100mg Medchemexpress, Vardenafil (hydrochloride) CAS:224785-91-5 Purity:>98%
SDP
Catalog No. 502082541
Encompass_Preferred
Medchemexpress, HY-B0442A 100mg Vardenafil (hydrochloride) CAS:224785-91-5 Vardenafil hydrochloride is a selective, orally active, potent inhibitor of phosphodiesterase-5 (PDE5), with an IC50 of 0.7 nM. Vardenafil hydrochloride shows selectivity over PDE1 (180 nM), PDE6 (11 nM), PDE2, PDE3, and PDE4 (>1000 nM). Vardenafil hydrochloride competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis and thus increases cGMP levels. Vardenafil hydrochloride can be used for the research of erectile dysfunction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2541 Supplier Medchemexpress LLC Supplier No. HYB0442A100MG
Medchemexpress, HY-B0442A 100mg Vardenafil (hydrochloride) CAS:224785-91-5 Vardenafil hydrochloride is a selective, orally active, potent inhibitor of phosphodiesterase-5 (PDE5), with an IC50 of 0.7 nM. Vardenafil hydrochloride shows selectivity over PDE1 (180 nM), PDE6 (11 nM), PDE2, PDE3, and PDE4 (>1000 nM). Vardenafil hydrochloride competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis and thus increases cGMP levels. Vardenafil hydrochloride can be used for the research of erectile dysfunction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.