Learn More
Medchemexpress LLC Vardenafil hydrochloride | 224785-91-5 | 99.8% | 1 ML
SDP
Catalog No. 5000407823
Encompass_Preferred
Vardenafil hydrochloride is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5), with an IC50 of 0.7 nM. It competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis, leading to increased cGMP levels and can be used for research related to erectile dysfunction, hepatitis, and diabetes.
  • Selective and orally active PDE5 inhibitor
  • Increases intracellular cGMP levels
  • Suitable for research on erectile dysfunction, hepatitis, and diabetes
Catalog No. 50-004-07823 Supplier Medchemexpress LLC Supplier No. HYB0442A10MM/1ML
Vardenafil hydrochloride is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5), with an IC50 of 0.7 nM. It competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis, leading to increased cGMP levels and can be used for research related to erectile dysfunction, hepatitis, and diabetes.
  • Selective and orally active PDE5 inhibitor
  • Increases intracellular cGMP levels
  • Suitable for research on erectile dysfunction, hepatitis, and diabetes