Learn More
Medchemexpress LLC Vardenafil (hydrochloride) | 224785-91-5 | 99.8% | 25 MG
SDP
Catalog No. 5000450975
Encompass_Preferred
Vardenafil hydrochloride is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5), with an IC50 of 0.7 nM. It competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis, thereby increasing cGMP levels.
  • Selective and orally active inhibitor of PDE5 (IC50 of 0.7 nM)
  • Inhibits PDE1 (IC50 of 180 nM) and PDE6 (IC50 of 11 nM)
  • Increases cGMP levels
  • Used in research for erectile dysfunction, hepatitis, and diabetes
Catalog No. 50-004-50975 Supplier Medchemexpress LLC Supplier No. 25MGHYB0442A
Vardenafil hydrochloride is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5), with an IC50 of 0.7 nM. It competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis, thereby increasing cGMP levels.
  • Selective and orally active inhibitor of PDE5 (IC50 of 0.7 nM)
  • Inhibits PDE1 (IC50 of 180 nM) and PDE6 (IC50 of 11 nM)
  • Increases cGMP levels
  • Used in research for erectile dysfunction, hepatitis, and diabetes