Learn More
Selleck Chemical LLC Vardenafil hydrochloride
SDP
Catalog No. 5000004093
Encompass_Preferred
Vardenafil (BAY38-9456 Levitra Staxyn) hydrochloride is a selective orally active potent inhibitor of phosphodiesterase (PDE) with IC50 of 0 7 nM 11 nM and 180 nM for PDE5 PDE6 and PDE1 respectively
Catalog No. 50-000-04093 Supplier Selleck Chemical LLC Supplier No. S595725MG
Vardenafil (BAY38-9456 Levitra Staxyn) hydrochloride is a selective orally active potent inhibitor of phosphodiesterase (PDE) with IC50 of 0 7 nM 11 nM and 180 nM for PDE5 PDE6 and PDE1 respectively