Learn More
Medchemexpress LLC Vardenafil hydrochloride trihydrate | 330808-88-3 | 99.7% | 200 MG
SDP
Catalog No. 5000406322
Encompass_Preferred
Vardenafil hydrochloride trihydrate is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5) with an IC50 of 0.7 nM. It shows inhibitory effects towards PDE1 and PDE6 with IC50s of 180 nM and 11 nM, respectively, while IC50s are >1000 nM for PDE3 and PDE4. It competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis, which increases cGMP levels. It can be used for research related to erectile dysfunction, hepatitis, and diabetes.
  • It is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5).
  • It has an IC50 of 0.7 nM for PDE5.
  • It inhibits PDE1 and PDE6 with IC50s of 180 nM and 11 nM.
  • It competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis.
  • It increases cGMP levels.
  • It can be used for the research of erectile dysfunction.
  • It can be used for the research of hepatitis.
  • It can be used for the research of diabetes.
Catalog No. 50-004-06322 Supplier Medchemexpress LLC Supplier No. HYB0442B200MG
Vardenafil hydrochloride trihydrate is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5) with an IC50 of 0.7 nM. It shows inhibitory effects towards PDE1 and PDE6 with IC50s of 180 nM and 11 nM, respectively, while IC50s are >1000 nM for PDE3 and PDE4. It competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis, which increases cGMP levels. It can be used for research related to erectile dysfunction, hepatitis, and diabetes.
  • It is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5).
  • It has an IC50 of 0.7 nM for PDE5.
  • It inhibits PDE1 and PDE6 with IC50s of 180 nM and 11 nM.
  • It competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis.
  • It increases cGMP levels.
  • It can be used for the research of erectile dysfunction.
  • It can be used for the research of hepatitis.
  • It can be used for the research of diabetes.