Learn More
Medchemexpress LLC Vardenafil (hydrochloride trihydrate) | 330808-88-3 | 99.7% | 25 MG
SDP
Catalog No. 5000450594
Encompass_Preferred
Vardenafil hydrochloride trihydrate is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5), with an IC50 of 0.7 nM. It competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis, thereby increasing cGMP levels. This compound can be used for research related to erectile dysfunction, hepatitis, and diabetes.
  • Selective and orally active inhibitor of phosphodiesterase-5 (PDE5).
  • Inhibits PDE1 and PDE6 with IC50s of 180 nM and 11 nM.
  • IC50s are >1000 nM for PDE3 and PDE4.
  • Increases intracellular cGMP levels.
  • Suitable for research of erectile dysfunction, hepatitis, and diabetes.
Catalog No. 50-004-50594 Supplier Medchemexpress LLC Supplier No. 25MGHYB0442B
Vardenafil hydrochloride trihydrate is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5), with an IC50 of 0.7 nM. It competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis, thereby increasing cGMP levels. This compound can be used for research related to erectile dysfunction, hepatitis, and diabetes.
  • Selective and orally active inhibitor of phosphodiesterase-5 (PDE5).
  • Inhibits PDE1 and PDE6 with IC50s of 180 nM and 11 nM.
  • IC50s are >1000 nM for PDE3 and PDE4.
  • Increases intracellular cGMP levels.
  • Suitable for research of erectile dysfunction, hepatitis, and diabetes.