Learn More
Medchemexpress LLC Vardenafil N-oxide | 448184-48-3 | >95.0% | C23H32N6O5S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13372410MG
Encompass_Preferred
Vardenafil N-oxide is the N-oxide impurity and analytical reference of the PDE5 inhibitor vardenafil, supplied for use in pharmaceutical analysis and research. It is used for impurity profiling, metabolite identification, and analytical method development, and is characterized by its CAS and molecular formula for unambiguous identification.
  • Analytical standard for impurity and metabolite identification.
  • Suitable for HPLC/UPLC method development and validation.
  • Provided in small, quantified amounts for reference use.
  • Molecular formula C23H32N6O5S and CAS 448184-48-3 for unambiguous identification.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-58770