Learn More
Cayman Chemical Vardenafil (hydrochloride hydrate), Purity: ≥98%, CAS: 330808-88-3, 50 mg
SDP
Catalog No. 502969517
Encompass_Preferred
An inhibitor of PDE5 (IC50s = 0.2-1.2 nM) and PDE6 (IC50 = 2 nM); selective for PDE5 and PDE6 over PDE1 and PDE11 (IC50s = 230 and 130 nM, respectively); improves erectile dysfunction in a rat model of acute arteriogenic erectile dysfunction; reduces the latency to mount and increases the frequency of mounting in mice after chronic treatment at 2 mg/kg for five weeks
Catalog No. 50-296-9517 Supplier Cayman Chemical Supplier No. 1493050 MG
An inhibitor of PDE5 (IC50s = 0.2-1.2 nM) and PDE6 (IC50 = 2 nM); selective for PDE5 and PDE6 over PDE1 and PDE11 (IC50s = 230 and 130 nM, respectively); improves erectile dysfunction in a rat model of acute arteriogenic erectile dysfunction; reduces the latency to mount and increases the frequency of mounting in mice after chronic treatment at 2 mg/kg for five weeks