Learn More
Cayman Chemical Varenicline-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2183239-01-0, 1 mg
SDP
Catalog No. 502936163
Encompass_Preferred
An internal standard for the quantification of varenicline by GC- or LC-MS
Catalog No. 50-293-6163 Supplier Cayman Chemical Supplier No. 261021 MG
An internal standard for the quantification of varenicline by GC- or LC-MS