Learn More
Cayman Chemical VarenIclIn-d4 500ug
SDP
Catalog No. 502936315
Encompass_Preferred
An internal standard for the quantification of varenicline by GC- or LC-MS
Catalog No. 50-293-6315 Supplier Cayman Chemical Supplier No. 26102500 UG
An internal standard for the quantification of varenicline by GC- or LC-MS