Learn More
Medchemexpress LLC Varenicline dihydrochloride | 866823-63-4 | 99.2% | 284.18 g/mol | C13H15Cl2N3 | 10 MG
SDP
Catalog No. 5000215237
Encompass_Preferred
Varenicline dihydrochloride is the dihydrochloride salt of varenicline, a selective α4β2 nicotinic acetylcholine receptor (nAChR) partial agonist. Supplied as a research-grade analytical standard for laboratory and in vitro studies, the product is supported by a data sheet, certificate of analysis, and safety data sheet. For research use only.
Catalog No. 50-002-15237 Supplier Medchemexpress LLC Supplier No. HY10019A10MG
Varenicline dihydrochloride is the dihydrochloride salt of varenicline, a selective α4β2 nicotinic acetylcholine receptor (nAChR) partial agonist. Supplied as a research-grade analytical standard for laboratory and in vitro studies, the product is supported by a data sheet, certificate of analysis, and safety data sheet. For research use only.