Learn More
Medchemexpress LLC Varenicline dihydrochloride | 866823-63-4 | 99.2% | 284.18 g/mol | C13H15Cl2N3 | 5 MG
SDP
Catalog No. 5000215236
Encompass_Preferred
Varenicline dihydrochloride is the dihydrochloride salt of a nicotinic acetylcholine receptor ligand used in research as a partial agonist at α4β2 receptors. Supplied as a solid powder for in vitro pharmacology, assay development, and reference standard applications, it offers consistent, well-characterized behavior for receptor-binding and functional studies.
  • High purity (99.2%) for research-grade use.
  • Dihydrochloride salt form for improved solubility in aqueous media.
  • Partial agonist at α4β2 nicotinic acetylcholine receptors, useful for studies of receptor pharmacology.
  • Known molecular weight (284.18 g/mol) and CAS number (866823-63-4) for unambiguous identification.
  • Supplied in small solid quantities suitable for assay development and reference standards.
Catalog No. 50-002-15236 Supplier Medchemexpress LLC Supplier No. HY10019A5MG
Varenicline dihydrochloride is the dihydrochloride salt of a nicotinic acetylcholine receptor ligand used in research as a partial agonist at α4β2 receptors. Supplied as a solid powder for in vitro pharmacology, assay development, and reference standard applications, it offers consistent, well-characterized behavior for receptor-binding and functional studies.
  • High purity (99.2%) for research-grade use.
  • Dihydrochloride salt form for improved solubility in aqueous media.
  • Partial agonist at α4β2 nicotinic acetylcholine receptors, useful for studies of receptor pharmacology.
  • Known molecular weight (284.18 g/mol) and CAS number (866823-63-4) for unambiguous identification.
  • Supplied in small solid quantities suitable for assay development and reference standards.