Learn More
Medchemexpress LLC Varenicline dihydrochloride | 866823-63-4 | 99.2% | 284.18 g/mol | C13H15Cl2N3 | 50 MG
SDP
Catalog No. 5000215723
Encompass_Preferred
Varenicline dihydrochloride is a small-molecule partial agonist of neuronal nicotinic acetylcholine receptors, used as a research reagent to study α4β2, α6β2, and α7 receptor pharmacology and mechanisms of nicotine dependence. The dihydrochloride salt improves aqueous solubility for in vitro assays and analytical workflows.
  • Potent partial agonist of α4β2 nicotinic receptors.
  • Also active at α6β2 and α7 receptor subtypes.
  • Suitable for in vitro pharmacology and signaling studies.
  • High reported purity appropriate for research use.
  • Dihydrochloride salt enhances solubility in aqueous media.
Catalog No. 50-002-15723 Supplier Medchemexpress LLC Supplier No. HY10019A50MG
Varenicline dihydrochloride is a small-molecule partial agonist of neuronal nicotinic acetylcholine receptors, used as a research reagent to study α4β2, α6β2, and α7 receptor pharmacology and mechanisms of nicotine dependence. The dihydrochloride salt improves aqueous solubility for in vitro assays and analytical workflows.
  • Potent partial agonist of α4β2 nicotinic receptors.
  • Also active at α6β2 and α7 receptor subtypes.
  • Suitable for in vitro pharmacology and signaling studies.
  • High reported purity appropriate for research use.
  • Dihydrochloride salt enhances solubility in aqueous media.