Learn More
Medchemexpress LLC HY-10020 50mg Medchemexpress, Varenicline (Hydrochloride) CAS:230615-23-3 Purity:>98%
SDP
Catalog No. 501931746
Encompass_Preferred
Medchemexpress, HY-10020 50mg Varenicline (Hydrochloride) CAS:230615-23-3 Varenicline Hydrochloride (CP 526555 hydrochloride) is a high affinity, selective α4β2 nicotine acetylcholine receptor (nAChR) partial agonist and full α7 nAChR agonist [1] [2] [3] . Varenicline Hydrochloride is also a potent partial agonist of α6β2 nAChR in striatum of rats with a K i value of 0.12 nM [4] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1746 Supplier Medchemexpress LLC Supplier No. HY1002050MG
Medchemexpress, HY-10020 50mg Varenicline (Hydrochloride) CAS:230615-23-3 Varenicline Hydrochloride (CP 526555 hydrochloride) is a high affinity, selective α4β2 nicotine acetylcholine receptor (nAChR) partial agonist and full α7 nAChR agonist [1] [2] [3] . Varenicline Hydrochloride is also a potent partial agonist of α6β2 nAChR in striatum of rats with a K i value of 0.12 nM [4] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.