Learn More
Medchemexpress LLC Varenicline hydrochloride | 230615-23-3 | 99.9% | 247.72 g/mol | C13H14ClN3 | 5 MG
SDP
Catalog No. 501931747
Encompass_Preferred
Varenicline hydrochloride is the hydrochloride salt of varenicline, an orally active partial agonist of the α4β2 nicotinic acetylcholine receptor. This compound is supplied as a research-grade chemical for in vitro and in vivo studies investigating nicotinic receptor pharmacology and models of nicotine dependence.
  • High purity suitable for analytical and preclinical studies.
  • Supplied as a hydrochloride salt for increased stability and solubility.
  • Molecular formula C13H14ClN3 and molecular weight 247.72 g/mol.
  • Identified by CAS number 230615-23-3 for unambiguous chemical reference.
  • Compatible with common solvents such as DMSO for solution preparation.
  • Intended for laboratory research use only.
Catalog No. 50-193-1747 Supplier Medchemexpress LLC Supplier No. HY100205MG
Varenicline hydrochloride is the hydrochloride salt of varenicline, an orally active partial agonist of the α4β2 nicotinic acetylcholine receptor. This compound is supplied as a research-grade chemical for in vitro and in vivo studies investigating nicotinic receptor pharmacology and models of nicotine dependence.
  • High purity suitable for analytical and preclinical studies.
  • Supplied as a hydrochloride salt for increased stability and solubility.
  • Molecular formula C13H14ClN3 and molecular weight 247.72 g/mol.
  • Identified by CAS number 230615-23-3 for unambiguous chemical reference.
  • Compatible with common solvents such as DMSO for solution preparation.
  • Intended for laboratory research use only.