Learn More
Selleck Chemical LLC Varenicline Hydrochloride S5022-25mg
SDP
Catalog No. 503112045
Encompass_Preferred
Varenicline (CP 526555) Hydrochloride is a partial agonist of 4 2 nicotine acetylcholine receptor (nAChR) and 6 2 nicotine acetylcholine receptor (nAChR) with a Ki value of 0 12 nM and 0 14 nM for 6 2 nAChR and 4 2 nAChR respectively in the striatum of rats
Catalog No. 50-311-2045 Supplier Selleck Chemical LLC Supplier No. S502225MG
Varenicline (CP 526555) Hydrochloride is a partial agonist of 4 2 nicotine acetylcholine receptor (nAChR) and 6 2 nicotine acetylcholine receptor (nAChR) with a Ki value of 0 12 nM and 0 14 nM for 6 2 nAChR and 4 2 nAChR respectively in the striatum of rats