Learn More
Medchemexpress LLC Varenicline hydrochloride | 230615-23-3 | 99.9% | 247.72 g/mol | C13H14ClN3 | 1 ML
SDP
Catalog No. 5000215680
Encompass_Preferred
Varenicline hydrochloride is a research-grade nicotinic acetylcholine receptor modulator used in neuroscience and pharmacology studies. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for receptor pharmacology, screening, and mechanistic experiments. CAS: 230615-23-3.
  • Partial agonist of α4β2 nicotinic acetylcholine receptors.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High purity suitable for research applications (99.94%).
  • Suitable for in vitro pharmacology and screening assays.
  • Recommended refrigerated storage and defined solvent stability.
Catalog No. 50-002-15680 Supplier Medchemexpress LLC Supplier No. HY1002010MM 1ML
Varenicline hydrochloride is a research-grade nicotinic acetylcholine receptor modulator used in neuroscience and pharmacology studies. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for receptor pharmacology, screening, and mechanistic experiments. CAS: 230615-23-3.
  • Partial agonist of α4β2 nicotinic acetylcholine receptors.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High purity suitable for research applications (99.94%).
  • Suitable for in vitro pharmacology and screening assays.
  • Recommended refrigerated storage and defined solvent stability.