Learn More
Medchemexpress LLC Varenicline hydrochloride | 230615-23-3 | 99.9% | 247.72 g/mol | C13H14ClN3 | 100 MG
SDP
Catalog No. 5000215875
Encompass_Preferred
Varenicline hydrochloride is the hydrochloride salt of varenicline, an orally active partial agonist of α4β2 nicotinic acetylcholine receptors used in neuroscience and pharmacology research. It is provided for research and analytical applications in solid form and as prepared solutions for in vitro studies and assay development.
  • Partial agonist at α4β2 nicotinic acetylcholine receptors, useful for receptor pharmacology studies.
  • Supplied as a powder and in solution formats for flexible assay preparation.
  • High purity suitable for analytical and biological assays.
  • Characterized with confirmed CAS number and molecular weight for chemical identification.
  • Suitable for use in studies of nicotine dependence and neuromodulation pathways.
Catalog No. 50-002-15875 Supplier Medchemexpress LLC Supplier No. HY10020100MG
Varenicline hydrochloride is the hydrochloride salt of varenicline, an orally active partial agonist of α4β2 nicotinic acetylcholine receptors used in neuroscience and pharmacology research. It is provided for research and analytical applications in solid form and as prepared solutions for in vitro studies and assay development.
  • Partial agonist at α4β2 nicotinic acetylcholine receptors, useful for receptor pharmacology studies.
  • Supplied as a powder and in solution formats for flexible assay preparation.
  • High purity suitable for analytical and biological assays.
  • Characterized with confirmed CAS number and molecular weight for chemical identification.
  • Suitable for use in studies of nicotine dependence and neuromodulation pathways.