Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Varenicline hydrochloride | 230615-23-3 | 99.9% | 247.72 g/mol | C13H14ClN3 | 25 MG
SDP

Catalog No. 5000215252
Encompass_Preferred

Varenicline hydrochloride is the hydrochloride salt of varenicline, an orally active partial agonist of the α4β2 nicotinic acetylcholine receptor. It is supplied as a research-grade chemical for research and analytical applications.

  • High purity (99.9%) suitable for research and analytical use.
  • Intended for research and analytical applications.
  • Available in multiple pack sizes, including 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • Molecular weight 247.72 g/mol and formula C13H14ClN3.
  • CAS number 230615-23-3 for unambiguous identification.

Catalog No. 50-002-15252 Supplier Medchemexpress LLC Supplier No. HY1002025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Varenicline hydrochloride is the hydrochloride salt of varenicline, an orally active partial agonist of the α4β2 nicotinic acetylcholine receptor. It is supplied as a research-grade chemical for research and analytical applications.

  • High purity (99.9%) suitable for research and analytical use.
  • Intended for research and analytical applications.
  • Available in multiple pack sizes, including 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • Molecular weight 247.72 g/mol and formula C13H14ClN3.
  • CAS number 230615-23-3 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.