Learn More
Selleck Chemical LLC Varenicline Tartrate (CP 526555-18) S1440-5mg
SDP
Catalog No. 503111318
Encompass_Preferred
Varenicline Tartrate (CP 526555-18 Chantix Champix) is a nicotinic AChR partial agonist used to treat nicotine addiction
Catalog No. 50-311-1318 Supplier Selleck Chemical LLC Supplier No. S14405MG
Varenicline Tartrate (CP 526555-18 Chantix Champix) is a nicotinic AChR partial agonist used to treat nicotine addiction