Learn More
Medchemexpress LLC Varenicline tartrate | 375815-87-5 | 100.0% | C17H19N3O6 | 1 ML
SDP
Catalog No. 5000215378
Encompass_Preferred
Varenicline tartrate is the tartrate salt of varenicline, a nicotinic acetylcholine receptor ligand used in research as a partial agonist of α4β2 nAChR. Supplied as a 10 mM solution in DMSO, this product is intended for laboratory research and analytical applications and is not for human use.
  • Ready-to-use 10 mM solution in DMSO.
  • High purity (99.96%) suitable for analytical applications.
  • Small 1 mL vial minimizes freeze-thaw and waste.
  • Validated for receptor pharmacology and in vitro assays.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-15378 Supplier Medchemexpress LLC Supplier No. HY1002110MM 1ML
Varenicline tartrate is the tartrate salt of varenicline, a nicotinic acetylcholine receptor ligand used in research as a partial agonist of α4β2 nAChR. Supplied as a 10 mM solution in DMSO, this product is intended for laboratory research and analytical applications and is not for human use.
  • Ready-to-use 10 mM solution in DMSO.
  • High purity (99.96%) suitable for analytical applications.
  • Small 1 mL vial minimizes freeze-thaw and waste.
  • Validated for receptor pharmacology and in vitro assays.
  • Stable when stored under recommended conditions.