Learn More
Medchemexpress LLC Varenicline tartrate | 375815-87-5 | MFCD08460603 | 100.0% | 361.127 Da | C17H19N3O6 | 25 MG
SDP
Catalog No. 5000215379
Encompass_Preferred
Varenicline tartrate is the tartrate salt of varenicline supplied as an analytical research standard for studies of nicotinic acetylcholine receptor pharmacology and nicotine dependence. It is an orally active partial agonist at α4β2 nicotinic acetylcholine receptors, provided as a high-purity solid suitable for analytical and pharmacological applications.
  • High purity suitable for analytical use.
  • Solid form for easy handling and storage.
  • Suitable for receptor pharmacology and nicotine dependence studies.
  • Provided as an analytical standard for quantitative assays.
  • Available in 25 mg pack sizes for small-scale research.
Catalog No. 50-002-15379 Supplier Medchemexpress LLC Supplier No. HY1002125MG
Varenicline tartrate is the tartrate salt of varenicline supplied as an analytical research standard for studies of nicotinic acetylcholine receptor pharmacology and nicotine dependence. It is an orally active partial agonist at α4β2 nicotinic acetylcholine receptors, provided as a high-purity solid suitable for analytical and pharmacological applications.
  • High purity suitable for analytical use.
  • Solid form for easy handling and storage.
  • Suitable for receptor pharmacology and nicotine dependence studies.
  • Provided as an analytical standard for quantitative assays.
  • Available in 25 mg pack sizes for small-scale research.