Learn More
Medchemexpress LLC Varenicline tartrate | 375815-87-5 | MFCD08460603 | >=98.0% | 361.3 g/mol | C13H13N3·C4H6O6 | 10 MG
SDP
Catalog No. 5000215587
Encompass_Preferred
Varenicline tartrate is the tartrate salt of varenicline, a partial agonist of α4β2 nicotinic acetylcholine receptors. Supplied as a research/analytical standard in solid form, it is intended for laboratory use such as pharmacology, method development, and analytical testing; not for human consumption.
  • High purity suitable for research and analytical applications (≥98% by HPLC).
  • Off-white to yellow crystalline solid.
  • Available in small vials, including a 10 MG pack size.
  • Provided as a defined chemical salt for reproducible assay results.
  • CAS number 375815-87-5 for unambiguous identification.
Catalog No. 50-002-15587 Supplier Medchemexpress LLC Supplier No. HY1002110MG
Varenicline tartrate is the tartrate salt of varenicline, a partial agonist of α4β2 nicotinic acetylcholine receptors. Supplied as a research/analytical standard in solid form, it is intended for laboratory use such as pharmacology, method development, and analytical testing; not for human consumption.
  • High purity suitable for research and analytical applications (≥98% by HPLC).
  • Off-white to yellow crystalline solid.
  • Available in small vials, including a 10 MG pack size.
  • Provided as a defined chemical salt for reproducible assay results.
  • CAS number 375815-87-5 for unambiguous identification.